Order Entry
France
Orders LinkContactUsLinkComponent
2-Iodoadenosine

2-Iodoadenosine

Bottle, 1 g
Catalog # APOSOR310610-1G
Supplier:   Apollo Scientific
2-Iodoadenosine
Catalog # APOSOR310610-1G
Supplier:  Apollo Scientific
CAS Number:  35109-88-7
2-Iodoadenosine

Bottle, 1 g

CAS Number:  35109-88-7
Price
94,10 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  (2R,3R,4S,5R)-2-(6-Amino-2-iodo-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol
  • CAS Number:35109-88-7
  • Formula:C₁₀H₁₂IN₅O₄
  • MDL Number:MFCD01320413
  • Molecular Weight:393.14 g/mol
  • Storage Temperature:Température ambiante
Specifications

Specifications

  • Type de conditionnement:
    Bottle
  • Cdt:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.