Order Entry
France
Orders LinkContactUsLinkComponent
1,4-Dioxane 99%

1,4-Dioxane 99%

Bottle, 1 L
Catalog # APOSOR61424-1L
Flammable
Irritant
Health
Supplier:   Apollo Scientific
1,4-Dioxane 99%
Catalog # APOSOR61424-1L
Supplier:  Apollo Scientific
CAS Number:  123-91-1
1,4-Dioxane 99%

Bottle, 1 L

CAS Number:  123-91-1
Price
37,70 €
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Glycolethylether, 1,4-Dioxacyclohexane, Dioxane, p-Dioxane, 1,4-Diethylene dioxide
  • CAS Number:123-91-1
  • Formula:C₄H₈O₂
  • MDL Number:MFCD00006571
  • Molecular Weight:88.11 g/mol
  • Boiling Pt:100…102 °C
  • Melting Pt:44540 °C
  • Density:1.034 g/cm³ (25 °C)
  • Flash Pt:12 °C
  • Storage Temperature:Ambient
  • UN:1165
  • ADR:3,II

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.