DUKE STANDARDS NIST TRACEABLE MEAN DIAME
Catalog # MSPPLTX137-15ML
Supplier: MARKET SOURCE PART PROCESS
DUKE STANDARDS NIST TRACEABLE MEAN DIAME
Catalog # MSPPLTX137-15ML
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Transfer pipettes
Supplier: RATIOLAB
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Pouring rings for laboratory bottles, DURAN®
Supplier: DWK Life Sciences
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Primary colour solutions, Reagent Ph. Eur.
Supplier: VWR Chemicals
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection