Order Entry
France
ContactUsLinkComponent
These tubes are ideal for glucose determinations. the vacuette® glucose tubes contain sodium fluoride as stabiliser with a choice of potassium oxalate, k3edta or sodium heparin as anticoagulant. vacuette® glucomedics tubes contain buffered na2-edta, 1 * 1.200 items
Catalog # GREI454033_1200
Supplier:  Greiner Bio-One
These tubes are ideal for glucose determinations. the vacuette® glucose tubes contain sodium fluoride as stabiliser with a choice of potassium oxalate, k3edta or sodium heparin as anticoagulant. vacuette® glucomedics tubes contain buffered na2-edta, 1 * 1.200 items
Catalog # GREI454033_1200
Supplier:  Greiner Bio-One
Frequently Bought Together