HPLC columns, Cogent C8 300™
Analytical, Bidentate C8, Stainless steel, 100 Å, 4 µm, 4,60 mm, 150 mm
Catalog # 554-6115
Supplier: MICROSOLV
Jump to:
Product Details & Documents
Using proprietary bonding technology, the Cogent C8 300™ has a C8 chain bonded directly to the 5 µm, 300A TYPE-C™ silica surface with two separate points of attachment resulting in silcon-carbon bonds.
- Extremely long column life
- Polar and non-polar analytes
- Increase precision of your results
- Based on silica hydride
- Extremely low pH is possible with this column
The bonded does not hydrolyse in acidic conditions since there are no siloxane bonds attaching the ligands. This column is a very stable, efficient and unique HPLC column for macro molecules. A very powerful tool for bio separations labs working with large proteins, peptides, polysaccharides or tryptic digests looking for longer column life and improved precision in results.
Specifications
- Column Format:Analytical
- Ø int.:4,60 mm
- Particle Size:4 µm
- Pore size:100 Å
- Length:150 mm
- Seq:24
- Column hardware:Stainless steel
- Phase:Bidentate C8
- Carbon load:7%
- Cat. No.:554-6115
- Endcapped:No
- pH range:2,0 - 10,1
- Surface area:350 m2/gm



