HPLC columns, Cogent C8 300™
Analytical, Bidentate C8 2.ō™, Stainless steel, 120 Å, 2,2 µm, 2,10 mm, 100 mm
Catalog # 554-6441
Supplier: MICROSOLV
Jump to:
Product Details & Documents
Using proprietary bonding technology, the Cogent C8 300™ has a C8 chain bonded directly to the 5 µm, 300A TYPE-C™ silica surface with two separate points of attachment resulting in silcon-carbon bonds.
- Extremely long column life
- Polar and non-polar analytes
- Increase precision of your results
- Based on silica hydride
- Extremely low pH is possible with this column
The bonded does not hydrolyse in acidic conditions since there are no siloxane bonds attaching the ligands. This column is a very stable, efficient and unique HPLC column for macro molecules. A very powerful tool for bio separations labs working with large proteins, peptides, polysaccharides or tryptic digests looking for longer column life and improved precision in results.
Specifications
- Column Format:Analytical
- Ø int.:2,10 mm
- Particle Size:2,2 µm
- Pore size:120 Å
- Length:100 mm
- Seq:77
- Column hardware:Stainless steel
- Phase:Bidentate C8 2.ō™
- Carbon load:11 - 12%
- Cat. No.:554-6441
- Endcapped:No
- pH range:2,0 - 8,0
- Surface area:340 m2/gm



