HPLC columns, Cogent C8 300™
Using proprietary bonding technology, the Cogent C8 300™ has a C8 chain bonded directly to the 5 µm, 300A TYPE-C™ silica surface with two separate points of attachment resulting in silcon-carbon bonds.
- Extremely long column life
- Polar and non-polar analytes
- Increase precision of your results
- Based on silica hydride
- Extremely low pH is possible with this column
The bonded does not hydrolyse in acidic conditions since there are no siloxane bonds attaching the ligands. This column is a very stable, efficient and unique HPLC column for macro molecules. A very powerful tool for bio separations labs working with large proteins, peptides, polysaccharides or tryptic digests looking for longer column life and improved precision in results.
- Column Format:Analytical
- Ø int.:2,10 mm
- Particle Size:2,2 µm
- Pore size:120 Å
- Length:50 mm
- Seq:75
- Column hardware:Stainless steel
- Phase:Bidentate C8 2.ō™
- Carbon load:11 - 12%
- Cat. No.:554-6437
- Endcapped:No
- pH range:2,0 - 8,0
- Surface area:340 m2/gm
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



