PreMoist foam tip swab in TSC Neutralising Buffer Foam tip swab offers enhanced collection and release of sample Will neutralise sanitisers and disinfectants White polystyrene breakpoint shaft Sample easily transferred to smaller vials Suitable for m 1 * 160 items
Catalog # TSCLTS/6-63BP
Supplier: TSC LIMITED
Frequently Bought Together