Order Entry
France
ContactUsLinkComponent
Mini temperature and humidity data logger for process monitoring. Tempmate B4 including 3-point calibration -20C to +85C Calibration at 0C, +25C and 50C 1 * 1 items
Catalog # IMEC100541
Supplier:  tempmate
Mini temperature and humidity data logger for process monitoring. Tempmate B4 including 3-point calibration -20C to +85C Calibration at 0C, +25C and 50C 1 * 1 items
Catalog # IMEC100541
Supplier:  tempmate
Frequently Bought Together