COLOUR STANDARD USP COBALTOUS CHLORIDE 1 * 100 mL
Catalog # REGEUSPCS103
Supplier: REAGECON
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Beakers, Tall Form
Supplier: VWR Collection
Sodium nitroprusside dihydrate 99.0-102.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Formamide, dry, Hydranal™ solubilizer for Karl Fischer titration, Fluka™
Supplier: Honeywell Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
pH buffers, ready to use
Supplier: WTW
VWR®, Primary colour solutions, Reagent Ph. Eur.
Supplier: VWR Chemicals
Trifluoroacetic acid 100%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Powder flasks with conical ground joint
Supplier: Lenz Laborglas GmbH & CO.KG