This gene encodes an enzyme that catalyzes the conversion of N-acetyl_L-aspartic acid (NAA) to aspartate and acetate. NAA is abundan t in the brain where hydrolysis by aspartoacylase is thought to help maintain white matter. 1 * 100 µG
Catalog # BWRLBS8799
Supplier: Bioworld Technology
Frequently Bought Together