SYMBOL: CA, MATRIX: HYDROCARBON OIL, SIZE: 50 G. METALLO-ORGANIC METALS IN HYDROCARBON OIL. ACCURACY ENSURED BY QUALITY TESTING WITH NIST STANDARD REFERENCE MATERIALS WHEN AVAILABLE. CERTIFICATE OF ANALYSIS SUPPLIED WITH EACH STANDARD. 1 * 1 items
Catalog # PERCN9308322
Supplier: PerkinElmer
Frequently Bought Together