SC 150-S14P (230V/50Hz) PLASTIC HEATING BATH CIRCULATOR, SC150-S14P, Amb+13 to 100 C, 8-14 L VOLUME, PLASTIC BATH, 2.0 kW HEATER, P UMP 17 L/MIN & 300 MBAR USB 1 * 1 items
Catalog # HAKA1533141
Supplier: THERMO ELECTRON LED
Frequently Bought Together