Order Entry
France
ContactUsLinkComponent
pH combination electrode, liquid filled, pH 0…14, -10…100°C 3x ceramic diaphragm, Glass body, 1 m cable, BNC + banana connector 1 * 1 items
Catalog # HACHLZW5015T97002
Supplier:  Hach
pH combination electrode, liquid filled, pH 0…14, -10…100°C 3x ceramic diaphragm, Glass body, 1 m cable, BNC + banana connector 1 * 1 items
Catalog # HACHLZW5015T97002
Supplier:  Hach
Frequently Bought Together