Cuvettes and holders
Glass flow through cell, 450 µl, 10 mm
Catalog # SCLI80-2004-45
Supplier: BIOCHROM
Jump to:
Product Details & Documents
Cuvettes for spectrophotometers designed to hold samples during spectroscopic experiments.
Different pathlengths and formats are available.
Specifications
- Path length:10 mm
- Volume:450 µl
- Description:Glass flow through cell
- Seq:02