Order Entry
France
Orders LinkContactUsLinkComponent
Cuvettes and holders
Cuvettes and holders
Programmable Peltier heated cell holder (20 - 105 °C)
Catalog # SCLI80-2106-14
Supplier:   BIOCHROM
Cuvettes and holders
Catalog # SCLI80-2106-14
Supplier:  BIOCHROM
Cuvettes and holders
Programmable Peltier heated cell holder (20 - 105 °C)
Price
Inquire For Price
Log in for availability
1 * 1 Pce
Product Details & Documents
Cuvettes for spectrophotometers designed to hold samples during spectroscopic experiments.
Different pathlengths and formats are available.
Specifications
  • Description:
    Programmable Peltier heated cell holder (20 - 105 °C)
  • Seq:
    09