Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
France
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Astrocap color: white size: 38cm description: latex free elastic material: polypropylene thickness: 20 g/m packaging: 100 pcs/bag, 1000pcs/carton 1 * 1.000 items
Catalog # GUNA140129
Supplier:
VWR Collection
Expand image
Astrocap color: white size: 38cm description: latex free elastic material: polypropylene thickness: 20 g/m packaging: 100 pcs/bag, 1000pcs/carton 1 * 1.000 items
Catalog # GUNA140129
Supplier:
VWR Collection
Frequently Bought Together
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
Nitrile gloves, TouchNTuff® 93-163 / 93-263
Supplier: Ansell
Nitrile gloves, TouchNTuff® 92-600 / 92-605
Supplier: Ansell
VWR® Nitrile Extra Light, Nitrile Gloves
Supplier: VWR Collection
VWR®, Latex Gloves
Supplier: VWR Collection
VWR® Spec-Wipe® 3, Cleanroom Wipes, Non Woven
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
Cleanroom gloves, polychloroprene, BioClean Fusion™
Supplier: Nitritex