A dual agonist of PPARα and γ (IC50s = 1 and 0.2 µM, respectively) that at 3 µM/kg/day for 3 weeks has been used to reduce insulin r esistance in obese Zucker rats 1 * 50 mg
Catalog # CAYM16791-50
Supplier: Cayman Chemical
Frequently Bought Together