Filament assembly, for Agilent 7010 Triple Quadrupole GC/MS 1 * 1 items
Catalog # HEWLG7002-60001
Supplier: Agilent
Filament assembly, for Agilent 7010 Triple Quadrupole GC/MS 1 * 1 items
Catalog # HEWLG7002-60001
Supplier: Agilent
Frequently Bought Together
Laboratory coats, howie style, C865
Supplier: Portwest
Dewar vessels, cylindrical, type C and G-C
Supplier: KGW
Flasks, flat bottom with standard ground joint
Supplier: Lenz Laborglas GmbH & CO.KG
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Mini Vortex Mixers
Supplier: VWR Collection
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
Storage bottles, octogonal, PET, Corning® Gosselin™
Supplier: Corning
Condenser clamp
Supplier: USBECK Laborgeräte
Dishes, disposable
Supplier: Korff
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
CABLE TIES NYLON NYTIE-3.5X200-NT 1 * 100 items
Supplier: Brady
Single-use syringes, 3-piece, HENKE-JECT®
Supplier: HENKE SASS WOLF