2-3-[4-(3-CHLORO-4-ETHYLPHENYL)PIPERAZIN-1-YL]PROPYL-1,2,4-TRIAZOLO[4,3-A]PYRIDIN-3(2H)-ONE HYDROCHLORIDE 1 * 50 mg
Catalog # PROOMM0421.14
Supplier: LGC Standards PROMOCHEM
2-3-[4-(3-CHLORO-4-ETHYLPHENYL)PIPERAZIN-1-YL]PROPYL-1,2,4-TRIAZOLO[4,3-A]PYRIDIN-3(2H)-ONE HYDROCHLORIDE 1 * 50 mg
Catalog # PROOMM0421.14
Supplier: LGC Standards PROMOCHEM
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection