Order Entry
France
Orders LinkContactUsLinkComponent
Ammonium chloride for analysis

Ammonium chloride for analysis

Glass bottle, 250 g
Catalog # ACRO393182500
Irritant
Ammonium chloride for analysis
Catalog # ACRO393182500
CAS Number:  12125-02-9
Ammonium chloride for analysis
Ammonium chloride for analysis

Glass bottle, 250 g

CAS Number:  12125-02-9
Price
43,10 €
Log in for availability
1 * 250 g
Product Details & Documents
Synonyms:  Ammonium muriate, Salmiac
  • CAS Number:12125-02-9
  • Formula:NH₄Cl
  • MDL Number:MFCD00011420
  • Molecular Weight:53.49 g/mol
  • Melting Pt:340 °C (subl.)
  • Density:1,5256 g/cm³ (20 °C)
  • Storage Temperature:Ambient

Documents

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder or crystals
  • Titration Argentometric:
    ≥99.0 %
  • Residue after ign. (sulfated):
    ≤0.01 %
  • Phosphate (PO4):
    ≤0.002 %
  • Sulfate (SO4):
    ≤0.002 %
  • Calcium (Ca):
    ≤0.001 %
  • Iron (Fe):
    ≤0.0005 %
  • Magnesium (Mg):
    ≤0.001 %
  • Heavy metals (as Pb):
    ≤10 ppm
  • Insoluble matter:
    ≤0.01 % (5 % in water)

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.