Ammonium chloride for analysis
Plastic drum, 5 kg
Supplier: Thermo Fisher Scientific
Ammonium chloride for analysis
Catalog # ACRO393180050
Supplier: Thermo Fisher Scientific
CAS Number: 12125-02-9
Jump to:
Product Details & Documents
Synonyms: Ammonium muriate, Salmiac
- CAS Number:12125-02-9
- Formula:NH₄Cl
- MDL Number:MFCD00011420
- Molecular Weight:53.49 g/mol
- Melting Pt:340 °C (subl.)
- Density:1,5256 g/cm³ (20 °C)
- Storage Temperature:Ambient
Documents
ACRO39318
SDS
Specifications
Specification Test Results
- Appearance:White Crystalline powder or crystals
- Titration Argentometric:≥99.0 %
- Residue after ign. (sulfated):≤0.01 %
- Phosphate (PO4):≤0.002 %
- Sulfate (SO4):≤0.002 %
- Calcium (Ca):≤0.001 %
- Iron (Fe):≤0.0005 %
- Magnesium (Mg):≤0.001 %
- Heavy metals (as Pb):≤10 ppm
- Insoluble matter:≤0.01 % (5 % in water)
Specifications
- Pack type:Plastic drum
- Pk:5 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



