Order Entry
France
Orders LinkContactUsLinkComponent
 

Crystal violet, certified

Glass bottle, 100 g
Catalog # ACRO405831000
Irritant
Health
Corrosive
Environment
Crystal violet, certified
Catalog # ACRO405831000
CAS Number:  548-62-9
Crystal violet, certified
Crystal violet, certified

Glass bottle, 100 g

CAS Number:  548-62-9
Price
110,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Methylrosanilide chloride, Basic violet 3, Viocid, N-(4-(Bis(4-(dimethylamino)phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-methylmethanamonium chloride, Violet crystal, Methyl violet 10B, Dimethyl((di(4-(dimethylamino)phenyl)methylen)-2,5-cyclohexadien-1-ylidene)ammonium chloride, CI 42555, 4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]dimethylammonium chloride, Hexamethyl violet, Hexamethyl-p-rosaniline chloride, Vianin, Gentian violet
  • CAS Number:548-62-9
  • Formula:C₂₅H₃₀ClN₃
  • MDL Number:MFCD00011750
  • Molecular Weight:407.99 g/mol
  • Merck Index:13,04406
  • Melting Pt:173 °C (dec.)
  • Density:1,19 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:208-953-6
  • UN:3077
  • ADR:9,III
Specifications

Specification Test Results

  • Appearance:
    Dark green Crystalline powder
  • Infrared spectrum:
    Conforms
  • Dye content:
    ≥88 %
  • UV Lambda max:
    589 to 594 nm
  • Absorption Ratio P-15/P+15:
    0.98 to 1.20
  • BSC Certification:
    Passes

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.