Sodium chloride, extra pure
Plastic bottle, 1 kg
Supplier: Thermo Fisher Scientific
Sodium chloride, extra pure
Catalog # ACRO194090010
Supplier: Thermo Fisher Scientific
CAS Number: 7647-14-5
Jump to:
Product Details & Documents
Synonyms: Saline, NaCl, Sea salt, Halite
- CAS Number:7647-14-5
- Formula:NaCl
- MDL Number:MFCD00003477
- Molecular Weight:58.44 g/mol
- Boiling Pt:1461 °C (760 mmHg)
- Melting Pt:801 °C
- Density:2,16 g/cm³ (20 °C)
- Storage Temperature:Ambient
Documents
Certificates
Search by Lot NumberACRO19409
SDS
ACRO194090010_TDS_EN ACRO194090250_TDS_EN
Product specification
Specifications
Specification Test Results
- Appearance:White Crystalline powder
- Titration Argentometric:≥99.0 %
- pH:5.0 to 8.0 (5 % in water at 20°C)
- Sulfate (SO4):≤100 ppm
- Barium (Ba):≤50 ppm
- Calcium (Ca):≤50 ppm
- Iron (Fe):≤50 ppm
- Potassium (K):≤100 ppm
- Heavy metals (as Pb):≤20 ppm
Specifications
- Pack type:Plastic bottle
- Pk:1 kg



