Order Entry
France
Orders LinkContactUsLinkComponent
 

Propionic acid 99+%, extra pure

Glass bottle, 2,5 L
Catalog # ACRO220130025
Corrosive
Flammable
Irritant
Propionic acid 99+%, extra pure
Catalog # ACRO220130025
CAS Number:  79-09-4
Propionic acid 99+%, extra pure
Propionic acid 99+%, extra pure

Glass bottle, 2,5 L

CAS Number:  79-09-4
Price
222,00 €
Log in for availability
1 * 2,5 L
Product Details & Documents
Synonyms:  Propanoic acid
  • CAS Number:79-09-4
  • Formula:C₃H₆O₂
  • MDL Number:MFCD00002756
  • Molecular Weight:74.08 g/mol
  • Merck Index:13,07917
  • Boiling Pt:141 °C (1013 hPa)
  • Melting Pt:–22 °C
  • Density:0.99 g/cm³
  • Flash Pt:51 °C
  • Storage Temperature:Ambient
  • EINECS:201-176-3
  • UN:3463
  • ADR:8,II
  • REACH:01-2119486971-24
Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3850 to 1.3870 (20°C, 589 nm)
  • Water:
    ≤0.1 %
  • GC:
    ≥99.0 %
  • Color scale:
    ≤10 APHA

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L

Specifications