Order Entry
France
Orders LinkContactUsLinkComponent
 

Potassium hydroxide ∼ 85%, pellets for analysis

Plastic drum, 10 kg
Catalog # ACRO134060100
Irritant
Corrosive
Potassium hydroxide ∼ 85%, pellets for analysis
Catalog # ACRO134060100
CAS Number:  1310-58-3
Potassium hydroxide ∼ 85%, pellets for analysis
Potassium hydroxide ∼ 85%, pellets for analysis

Plastic drum, 10 kg

CAS Number:  1310-58-3
Price
486,00 €
Log in for availability
1 * 10 kg
Product Details & Documents
Synonyms:  Potash caustic, Caustic potash
  • CAS Number:1310-58-3
  • Formula:KOH
  • MDL Number:MFCD00003553
  • Molecular Weight:56.11 g/mol
  • Merck Index:12,07806
  • Boiling Pt:1320 °C (760 mmHg)
  • Melting Pt:360 °C
  • Density:2,04 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-181-3
  • UN:1813
  • ADR:8,II
  • REACH:01-2119487136-33
Specifications

Specification Test Results

  • Appearance:
    White Pellets
  • Titration with HCl:
    ≥85 %
  • Chloride (Cl):
    ≤70 ppm
  • Phosphate (PO4):
    ≤4 ppm
  • Sulfate (SO4):
    ≤10 ppm
  • Potassium carbonate:
    ≤1 % (K2CO3)
  • Aluminium (Al):
    ≤10 ppm
  • Arsenic (As):
    ≤0.5 ppm
  • Calcium (Ca):
    ≤10 ppm
  • Iron (Fe):
    ≤5 ppm
  • Nickel (Ni):
    ≤3 ppm
  • Silver (Ag):
    ≤0.5 ppm
  • Sodium (Na):
    ≤0.4 %
  • Heavy metals (as Pb):
    ≤10 ppm
  • Silicon dioxide (SiO2):
    ≤5 ppm
  • Nitrogen compounds (as N):
    ≤5 ppm

Specifications

  • Pack type:
    Plastic drum
  • Pk:
    10 kg

Specifications