Aerosol Filter Pipet Tips 200uL Low Retention. Rnase/Dnase, Endotoxin, DNA free. Tips fit most 200ul pipettors. In hinged rack. Tips are autoclavable. 96 tips per rack, 10 racks per pack. 10 packs per case. 9600 tips per case. Sterile 1 * 1 SET
Catalog # VWRI732-1489BOM
Supplier: VWR Collection
Frequently Bought Together