Flasks, round bottom, with standard ground joints
50 ml, 14/23 NS, 90 mm, 51 mm
WITG0.600.050.E
Borosilicate glass 3.3.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
One centre neck, useful for distillation and extraction.
- Ext.Ø:51 mm
- Socket size:14/23 NS
- Capacity:50 ml
- Height:90 mm
- Seq:09



