Flasks, round bottom, with standard ground joints
100 ml, 19/26 NS, 110 mm, 64 mm
WITG0.601.100.E
Borosilicate glass 3.3.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
One centre neck, useful for distillation and extraction.
- Ext.Ø:64 mm
- Socket size:19/26 NS
- Capacity:100 ml
- Height:110 mm
- Seq:14
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



