Order Entry
France
ContactUsLinkComponent
Mode of action and Anti-microbial spectrum: Ampicillin is an inhibitor of bacterial cell wall synthesis which acts by inhibiting the cross-linking of peptidoglycan. It is used in the selection of antibiotic cell hybrids, and has a high rate of actio 1 * 500 g/mol
Catalog # GENORC-157
Supplier:  G-Biosciences
Mode of action and Anti-microbial spectrum: Ampicillin is an inhibitor of bacterial cell wall synthesis which acts by inhibiting the cross-linking of peptidoglycan. It is used in the selection of antibiotic cell hybrids, and has a high rate of actio 1 * 500 g/mol
Catalog # GENORC-157
Supplier:  G-Biosciences
Frequently Bought Together