Crystal violet solution
3×1 l, 3
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Primary stain for Gram staining
Synonyms: Methylrosanilide chloride, Basic violet 3, Viocid, N-(4-(Bis(4-(dimethylamino)phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-methylmethanamonium chloride, Violet crystal, Methyl violet 10B, Dimethyl((di(4-(dimethylamino)phenyl)methylen)-2,5-cyclohexadien-1-ylidene)ammonium chloride, CI 42555, 4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]dimethylammonium chloride, Hexamethyl violet, Hexamethyl-p-rosaniline chloride, Vianin, Gentian violet
- CAS Number:548-62-9
- Formula:C₂₅H₃₀ClN₃
- MDL Number:MFCD00011750
- Molecular Weight:407.99 g/mol
Documents
Certificates
Search by Lot Number911517ZA_VIOLET CRYSTAL SOLUTION_ISO 11133_rev1.pdf 911517ZAMP_VIOLET CRYSTAL SOLUTION_ISO 11133_rev1.pdf
spec_VWRC911517ZA
Product specification
VWR Microbiology
Product Literature
Specifications
Specifications
- Pk:3
- Pk Info:3×1 l



