Order Entry
France
Orders LinkContactUsLinkComponent
Trays
Trays
Test tube tray, compatible with SR test tube racks or can be used alone to accommodate bags and miscellaneous vessels, for LSB12 (holds up to 3× SR test tube racks)
Catalog # 462-0540
Supplier:   GRANT INSTRUMENTS
Trays
Catalog # 462-0540
Supplier:  GRANT INSTRUMENTS
Trays
Trays
Test tube tray, compatible with SR test tube racks or can be used alone to accommodate bags and miscellaneous vessels, for LSB12 (holds up to 3× SR test tube racks)
Price
386,00 €
Log in for availability
1 * 1 Pce
Product Details & Documents
Specifications
  • Description:
    Test tube tray, compatible with SR test tube racks or can be used alone to accommodate bags and miscellaneous vessels, for LSB12 (holds up to 3× SR test tube racks)
  • Seq:
    04