Order Entry
France
Orders LinkContactUsLinkComponent
Trays
Trays
Test tube tray, compatible with SR test tube racks or can be used alone to accommodate bags and miscellaneous vessels, for OLS26 (holds up to 5× SR test tube racks)
Catalog # 462-0542
Supplier:   GRANT INSTRUMENTS
Trays
Catalog # 462-0542
Supplier:  GRANT INSTRUMENTS
Trays
Trays
Test tube tray, compatible with SR test tube racks or can be used alone to accommodate bags and miscellaneous vessels, for OLS26 (holds up to 5× SR test tube racks)
Price
409,00 €
Log in for availability
1 * 1 Pce
Product Details & Documents
Specifications
  • Description:
    Test tube tray, compatible with SR test tube racks or can be used alone to accommodate bags and miscellaneous vessels, for OLS26 (holds up to 5× SR test tube racks)
  • Seq:
    06