qPCR detection of West Nile Virus. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, Endoge nous control, extraction control 1 * 150 Tests
Catalog # Z-PATH-WNV
Supplier: PRIMERDESIGN
Frequently Bought Together