Order Entry
France
ContactUsLinkComponent
qPCR detection of Avian adenovirus EDS76 Egg Drop Syndrome. Contains: Primer and probe mix, Reverse Transcription target specific pr imers (RNA genome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-EDS-S
Supplier:  PRIMERDESIGN
qPCR detection of Avian adenovirus EDS76 Egg Drop Syndrome. Contains: Primer and probe mix, Reverse Transcription target specific pr imers (RNA genome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-EDS-S
Supplier:  PRIMERDESIGN
Frequently Bought Together