Order Entry
France
ContactUsLinkComponent
qPCR detection of Escherichia coli 0157:H7. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genom e viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-E-CH7
Supplier:  PRIMERDESIGN
qPCR detection of Escherichia coli 0157:H7. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genom e viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-E-CH7
Supplier:  PRIMERDESIGN
Frequently Bought Together