Order Entry
France
ContactUsLinkComponent
qPCR detection of shiga toxin (stx1) producing Escherichia coli. Contains: Primer and probe mix, Reverse Transcription target specif ic primers (RNA genome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-E.CO1
Supplier:  PRIMERDESIGN
qPCR detection of shiga toxin (stx1) producing Escherichia coli. Contains: Primer and probe mix, Reverse Transcription target specif ic primers (RNA genome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-E.CO1
Supplier:  PRIMERDESIGN
Frequently Bought Together