Order Entry
France
ContactUsLinkComponent
qPCR detection of Human Metapneumovirus. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genome v iruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-HMPV-
Supplier:  PRIMERDESIGN
qPCR detection of Human Metapneumovirus. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genome v iruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-HMPV-
Supplier:  PRIMERDESIGN
Frequently Bought Together