Order Entry
France
ContactUsLinkComponent
qPCR detection of Coronavirus 2012 genomes. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genom e viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-HCOV
Supplier:  PRIMERDESIGN
qPCR detection of Coronavirus 2012 genomes. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genom e viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-HCOV
Supplier:  PRIMERDESIGN
Frequently Bought Together