Ref gene selection kit human. 12 most stable genes (primer ony) plus qbase+ licence 1 * 1 items
Catalog # GEPLUS-SY-12-H
Supplier: PRIMERDESIGN
Ref gene selection kit human. 12 most stable genes (primer ony) plus qbase+ licence 1 * 1 items
Catalog # GEPLUS-SY-12-H
Supplier: PRIMERDESIGN
Frequently Bought Together
Decontamination solution, RNase AWAY®
Supplier: Thermo Fisher Scientific
<i>Tritirachium album</i> proteinase K
Supplier: PanReac AppliChem
VWR®, Breatheable Rayon Film Seals for Biological Cultures
Supplier: VWR Collection
Tris-Acetate-EDTA (TAE) buffer, 50X
Supplier: PanReac AppliChem
Agarose (100 bp - 23 kb), Electran, DNA grade for electrophoresis
Supplier: VWR Chemicals
Duo-Ready DNA Ultra 2.0 prefilled plates, KingFisher™
Supplier: Thermo Fisher Scientific
DNA ScreenTape Analysis (ScreenTape and Reagents)
Supplier: AGILENT
High Sensitivity DNA ScreenTape Analysis (ScreenTape and Reagents)
Supplier: AGILENT
Master mixes, DreamTaq™ Hot Start PCR master mix (2X)
Supplier: Thermo Fisher Scientific
Incucyte® Annexin V Dye for Apoptosis
Supplier: Sartorius
EasySep™ Buffer
Supplier: STEMCELL Technologies
Bovine desoxyribonuclease I (from Pancreas)
Supplier: PanReac AppliChem
Mag-Bind® FFPE DNA/RNA 96 Kit
Supplier: OMEGA BIO-TEK