Order Entry
France
ContactUsLinkComponent
qPCR detection of Respiratory Syncytial Virus A. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, Endogenous control, extraction control 1 * 150 Tests
Catalog # Z-PATH-RSV-A
Supplier:  PRIMERDESIGN
qPCR detection of Respiratory Syncytial Virus A. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, Endogenous control, extraction control 1 * 150 Tests
Catalog # Z-PATH-RSV-A
Supplier:  PRIMERDESIGN
Frequently Bought Together