Order Entry
France
ContactUsLinkComponent
qPCR detection of Moraxella catarrhalis. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-M.CATARRH
Supplier:  PRIMERDESIGN
qPCR detection of Moraxella catarrhalis. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-M.CATARRH
Supplier:  PRIMERDESIGN
Frequently Bought Together