Order Entry
France
ContactUsLinkComponent
qPCR detection of Chikungunya Virus. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, Endo genous control, extraction control 1 * 150 Tests
Catalog # Z-PATH-CHIKV
Supplier:  PRIMERDESIGN
qPCR detection of Chikungunya Virus. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, Endo genous control, extraction control 1 * 150 Tests
Catalog # Z-PATH-CHIKV
Supplier:  PRIMERDESIGN
Frequently Bought Together