Order Entry
France
ContactUsLinkComponent
Real-time PCR Genotyping kit to detect GSTP1 codon 105. Contains: Primer and multiplex probe mix - wildtype/variants, Positive contr ols - wildtype/variants 1 * 50 Tests
Catalog # PATH-GT-GSTP1A
Supplier:  PRIMERDESIGN
Real-time PCR Genotyping kit to detect GSTP1 codon 105. Contains: Primer and multiplex probe mix - wildtype/variants, Positive contr ols - wildtype/variants 1 * 50 Tests
Catalog # PATH-GT-GSTP1A
Supplier:  PRIMERDESIGN
Frequently Bought Together