Order Entry
France
ContactUsLinkComponent
qPCR detection of Sudan Ebola Virus. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, Endo genous control, extraction control 1 * 150 Tests
Catalog # PATH-SUDV
Supplier:  PRIMERDESIGN
qPCR detection of Sudan Ebola Virus. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, Endo genous control, extraction control 1 * 150 Tests
Catalog # PATH-SUDV
Supplier:  PRIMERDESIGN
Frequently Bought Together