Order Entry
France
ContactUsLinkComponent
Disposable Funnel made of High Density Polyethylene (HDPE) - virgin. Not Pre-Sterilised. Conforms to FDA CFR 177.1520. Conforms to EU Regs 10/2011. Overall Diameter: 195mm. Height: 190mm. Nominal Weight: 91g. Assembly & Packing Environment: Class 10
Catalog # SAMP8370H-200_U
Supplier:  Sampling Systems
Disposable Funnel made of High Density Polyethylene (HDPE) - virgin. Not Pre-Sterilised. Conforms to FDA CFR 177.1520. Conforms to EU Regs 10/2011. Overall Diameter: 195mm. Height: 190mm. Nominal Weight: 91g. Assembly & Packing Environment: Class 10
Catalog # SAMP8370H-200_U
Supplier:  Sampling Systems