Order Entry
France
ContactUsLinkComponent
C18G x 5µm particle size x 300Å pore size, 100mm length x 150µm ID, PEEKsil ProteCol HPLC Capillary Column 1 * 1 items
Catalog # SGEA2C184-05M15K
C18G x 5µm particle size x 300Å pore size, 100mm length x 150µm ID, PEEKsil ProteCol HPLC Capillary Column 1 * 1 items
Catalog # SGEA2C184-05M15K
Frequently Bought Together