Order Entry
France
ContactUsLinkComponent
C18H x 3µm particle size x 120Å pore size, 50mm length x 2.1mm ID, PEEK-coated ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2C182-03L02P
C18H x 3µm particle size x 120Å pore size, 50mm length x 2.1mm ID, PEEK-coated ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2C182-03L02P
Frequently Bought Together