Order Entry
France
ContactUsLinkComponent
C8H x 3µm particle size x 120Å pore size, 150mm length x 3.0mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2C83-03N03S
C8H x 3µm particle size x 120Å pore size, 150mm length x 3.0mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2C83-03N03S
Frequently Bought Together