Order Entry
France
Orders LinkContactUsLinkComponent
Grounding cables, Super Erecta®
Grounding cables, Super Erecta®
Grounding cable with universal bracket
Catalog # METOGCB16SA
Supplier:   Metro International
Grounding cables, Super Erecta®
Catalog # METOGCB16SA
Grounding cables, Super Erecta®
Grounding cables, Super Erecta®
Grounding cable with universal bracket
Price
107,00 €
Log in for availability
1 * 1 Pce
Product Details & Documents
These grounding cables provide an effective method of electrostatic discharge for any Metro Super Erecta unit (stationary or mobile) when used in conjunction with aluminum split sleeves or ESD kit and conductive casters.
  • Cables ensure a proper path to ground when used with ESD flooring
  • Grounding cable with universal bracket fits wire and solid shelves, as well as open-wire frames
Specifications
  • Description:
    Grounding cable with universal bracket
  • Seq:
    02